Skip to Content
Merck
All Photos(1)

Key Documents

510424

Sigma-Aldrich

3-Butylthiophene

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$49.20
5 G
$193.00

$49.20


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$49.20
5 G
$193.00

About This Item

Empirical Formula (Hill Notation):
C8H12S
CAS Number:
Molecular Weight:
140.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$49.20


0 left in stock

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.501 (lit.)

bp

181-183 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

SMILES string

CCCCc1ccsc1

InChI

1S/C8H12S/c1-2-3-4-8-5-6-9-7-8/h5-7H,2-4H2,1H3

InChI key

KPOCSQCZXMATFR-UHFFFAOYSA-N

General description

3-Butylthiophene is an alkylthiophene.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service