Skip to Content
Merck
All Photos(1)

Key Documents

488097

Sigma-Aldrich

Ammonium-15N2 sulfate

60 atom % 15N

Synonym(s):

15N Labeled ammonium sulfate, Diazanium-15N2 sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$874.00

$874.00


Check Cart for Availability

Request more information

Select a Size

Change View
5 G
$874.00

About This Item

Linear Formula:
(15NH4)2SO4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

$874.00


Check Cart for Availability

Request more information

isotopic purity

60 atom % 15N

Quality Level

mol wt

133.34 by atom % calculation (60 atom % 15N)

mp

>280 °C (dec.) (lit.)

application(s)

agriculture

mass shift

M+2

SMILES string

[15NH3].[15NH3].OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+1;

InChI key

BFNBIHQBYMNNAN-ISOLYIDJSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Tracer techniques using isotopic N have been employed since the 1940s to investigate such topics as N transformations and cycling in agricultural and other soils, N sources for crop uptake, the fate and behavior of fertilizer N applied to soils, the nature and extent of soil and fertilizer losses, and fertilizer N uptake efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service