Skip to Content
Merck
All Photos(1)

Key Documents

479179

Sigma-Aldrich

(S)-(−)-4-Hydroxy-2-pyrrolidinone

97%

Synonym(s):

(S)-β-Hydroxy-γ-butyrolactam

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$381.00

$381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$381.00

About This Item

Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
Beilstein:
1524192
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$381.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

optical activity

[α]23/D −43°, c = 1 in ethanol

mp

156-159 °C (lit.)

functional group

hydroxyl

SMILES string

O[C@@H]1CNC(=O)C1

InChI

1S/C4H7NO2/c6-3-1-4(7)5-2-3/h3,6H,1-2H2,(H,5,7)/t3-/m0/s1

InChI key

IOGISYQVOGVIEU-VKHMYHEASA-N

General description

4-Hydroxy-2-pyrrolidinone is an important building block found in many bioactive compounds like streptopyrrolidine. It can be used as an intermediate in the synthesis of various γ-amino acids (GABA) and substituted 2-pyrrolidinones like cynometrine and cynodine.[1]

Application

(S)-(−)-4-Hydroxy-2-pyrrolidinone can be used as a starting material in the preparation of:
  • Biologically significant pyrrolo[1,2:1′,2′]azepino[5,6-b]indole derivatives.[2]
  • Substituted azepanes by reacting with various aldehydes via photochemical [5+2] cycloaddition.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Streptopyrrolidine, an angiogenesis inhibitor from a marine-derived Streptomyces sp. KORDI-3973
Shin HJ, et al.
Phytochemistry, 69(12), 2363-2366 (2008)
A Photochemical Two-Step Formal [5+ 2] Cycloaddition: A Condensation-Ring-Expansion Approach to Substituted Azepanes
Thullen SM, et al.
Synlett, 28(20), 2755-2758 (2017)
First synthesis of pyrrolo [1, 2: 1′, 2′] azepino [5, 6-b] indole derivatives
Perron J, et al.
Tetrahedron Letters, 44(35), 6553-6556 (2003)
G Di Silvestro et al.
Journal of pharmaceutical sciences, 82(7), 758-760 (1993-07-01)
The phase diagram of (R)- and (S)-4-hydroxy-2-pyrrolidone presents a conglomerate in the racemic mixture. The enthalpy of melting extrapolated by the Schröder-van Laar-Le Chatelier equation [change in enthalpy (delta H) = 28410 J/mol; melting temperature (TA) = 429.9 K; solidus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service