Merck
  • Phase diagram of (R)- and (S)-4-hydroxy-2-pyrrolidone mixtures: a new case of a conglomerate-forming system.
Phase diagram of (R)- and (S)-4-hydroxy-2-pyrrolidone mixtures: a new case of a conglomerate-forming system.
Journal of pharmaceutical sciences (1993-07-01)
G Di Silvestro, G Palmisano, R Pellegata
ABSTRACT
The phase diagram of (R)- and (S)-4-hydroxy-2-pyrrolidone presents a conglomerate in the racemic mixture. The enthalpy of melting extrapolated by the Schröder-van Laar-Le Chatelier equation [change in enthalpy (delta H) = 28410 J/mol; melting temperature (TA) = 429.9 K; solidus temperature (Ts) = 395.4 K] and the value determined by differential scanning calorimetry (delta H = 28494 J/mol; TA = 429.6 K; Ts = 394.6 K) are in excellent agreement. The experimental entropy of the mixing of enantiomers is 5.69 J/mol.K. The conglomerate nature of the racemic mixture was confirmed by X-ray and IR spectroscopy. The presence of solvates was also observed.
Sigma-Aldrich
(R)-(+)-4-Hydroxy-2-pyrrolidinone, 97%
Sigma-Aldrich
(S)-(−)-4-Hydroxy-2-pyrrolidinone, 97%