Skip to Content
Merck
All Photos(1)

Documents

459941

Sigma-Aldrich

(R)-(−)-Benzoin

98%

Synonym(s):

(R)-2-Hydroxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

98%

optical activity

[α]24/D −115°, c = 1.5 in acetone

optical purity

ee: 99% (HPLC)

mp

135-137 °C (lit.)

SMILES string

O[C@H](c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H/t13-/m1/s1

InChI key

ISAOCJYIOMOJEB-CYBMUJFWSA-N

Application

(R)-(-)-Benzoin may be used in the preparation of (R)-2-hydroxy-1-phenylpropanone by reacting with benzaldehyde lyase (BAL) in the presence of acetaldehyde.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hung-Wei Tsui et al.
Journal of chromatography. A, 1637, 461796-461796 (2021-01-03)
The effect of solvents on the enantioselectivities of four structurally similar chiral solutes with a cellulose derivative-based chiral stationary phase, Chiralpak IB, were studied using acetone (AC), 2-propanol (IPA), and tert-butanol (TBA) separately as polar modifiers. The enantioselectivities α of
Rüdiger Ohs et al.
Biotechnology progress, 35(6), e2868-e2868 (2019-06-18)
The kinetic description of enzyme-catalyzed reactions is a core task in biotechnology and biochemical engineering. In particular, mechanistic kinetic models help from the discovery of the biocatalyst throughout its application. Chemo- or enantioselective enzyme reactions often undergo two alternative pathways
Rüdiger Ohs et al.
Biotechnology progress, 34(5), 1081-1092 (2018-06-10)
Thiamine diphosphate (ThDP)-dependent enzymes catalyze a broad range of reactions with excellent enantioselectivity. Among these reactions, carboligations of aldehydes are of particular interest since the products, chiral hydroxy ketones, are valuable building blocks in the pharmaceutical industry. However, the substrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service