Skip to Content
Merck
All Photos(2)

Key Documents

410128

Sigma-Aldrich

Scandium(III) isopropoxide

Synonym(s):

Scandium triisopropoxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$305.00

$305.00


Estimated to ship on20 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
$305.00

About This Item

Linear Formula:
Sc(OCH(CH3)2)3
CAS Number:
Molecular Weight:
222.22
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$305.00


Estimated to ship on20 April 2025


Request a Bulk Order

Quality Level

form

powder or granules
solid

composition

Sc, 19.5-22.0% EDTA titration

reaction suitability

core: scandium
reagent type: catalyst

SMILES string

CC(C)O[Sc](OC(C)C)OC(C)C

InChI

1S/3C3H7O.Sc/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

SLACPSWSQQIQTG-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service