Skip to Content
Merck
All Photos(1)

Documents

456691

Sigma-Aldrich

Disperse Black 9

Dye content 97 %

Synonym(s):

2,2′-[4-(4-Aminophenylazo)phenylimino]diethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4N=NC6H4N(CH2CH2OH)2
CAS Number:
Molecular Weight:
300.36
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 97%

mp

157-160 °C (lit.)

λmax

461 nm

ε (extinction coefficient)

≥20000 at 458-464 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc(cc1)N=Nc2ccc(cc2)N(CCO)CCO

InChI

1S/C16H20N4O2/c17-13-1-3-14(4-2-13)18-19-15-5-7-16(8-6-15)20(9-11-21)10-12-22/h1-8,21-22H,9-12,17H2

InChI key

NZKTVPCPQIEVQT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service