Skip to Content
Merck
All Photos(1)

Key Documents

454486

Sigma-Aldrich

(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde

Synonym(s):

(R)-(+)-Glyceraldehyde acetonide, (R)-Isopropylideneglyceraldehyde, NSC 89869

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$117.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$117.00

About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

optical activity

[α]22/D +53.8°, c = 2 in chloroform

Quality Level

refractive index

n20/D 1.454 (lit.)

bp

139 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

functional group

aldehyde
ether
ketal

storage temp.

−20°C

SMILES string

CC1(C)OC[C@@H](O1)C=O

InChI

1S/C6H10O3/c1-6(2)8-4-5(3-7)9-6/h3,5H,4H2,1-2H3/t5-/m0/s1

InChI key

YSGPYVWACGYQDJ-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in organic reactions including:
  • Henry condensation reactions
  • Synthesis of highly functionalized chiral aziridines
  • Olefination of aldehydes
  • C-H oxidations
  • Aldol-type condensations
  • Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents
  • Hosomi-Sakurai reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service