Skip to Content
Merck
All Photos(2)

Key Documents

411361

Sigma-Aldrich

Diethyl (bromodifluoromethyl)phosphonate

96%

Synonym(s):

(Bromodifluoromethyl)phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$121.00

$121.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$121.00

About This Item

Linear Formula:
BrCF2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
267.01
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$121.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

96%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.417 (lit.)

bp

40-41 °C/0.05 mmHg (lit.)

density

1.503 g/mL at 25 °C (lit.)

functional group

bromo
fluoro
phosphonate

SMILES string

CCOP(=O)(OCC)C(F)(F)Br

InChI

1S/C5H10BrF2O3P/c1-3-10-12(9,11-4-2)5(6,7)8/h3-4H2,1-2H3

InChI key

QRADKVYIJIAENZ-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3)
  • Synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps
  • P-C bond cleavage on basic hydrolysis
  • Preparation of a prodrug for intracellular delivery of protein tyrosine phosphatase 1B (PTP1B) inhibitor (nonhydrolyzable difluoromethylphosphonate moiety)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service