Skip to Content
Merck
All Photos(1)

Key Documents

447455

Sigma-Aldrich

Methyl 10-bromodecanoate

90%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)9CO2CH3
CAS Number:
Molecular Weight:
265.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

refractive index

n20/D 1.464 (lit.)

bp

186 °C (lit.)

density

1.137 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCCCCCCCBr

InChI

1S/C11H21BrO2/c1-14-11(13)9-7-5-3-2-4-6-8-10-12/h2-10H2,1H3

InChI key

SHCRSWJVWSAMKQ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of alkynylcyclooctatetraenes and alkynylcubanes.
Eaton PE and Stossel D.
The Journal of Organic Chemistry, 56(17), 5138-5142 (1991)
P Lu et al.
Archives of biochemistry and biophysics, 337(1), 1-7 (1997-01-01)
A series of omega-substituted fatty acids with potential heme-coordinating groups was synthesized as inhibitors of lauric acid omega-hydroxylation. The compounds were evaluated using liver microsomes from clofibrate (CF)-induced rats and an engineered expressed CYP4A1-derived fusion protein called f4A1. omega-Imidazolyl-decanoic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service