Skip to Content
Merck
All Photos(2)

Key Documents

443042

Sigma-Aldrich

2-Benzothiazoleacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2S
CAS Number:
Molecular Weight:
174.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

98-101 °C (lit.)

SMILES string

N#CCc1nc2ccccc2s1

InChI

1S/C9H6N2S/c10-6-5-9-11-7-3-1-2-4-8(7)12-9/h1-4H,5H2

InChI key

ZMZSYUSDGRJZNT-UHFFFAOYSA-N

Application

2-Benzothiazoleacetonitrile may be used for the preparation of 3-aryl-1-[(E)-cyanomethylidene]-1H-pyrido[2,1-b]benzothiazole-4-carbonitriles, via reaction with with 6-aryl-4-methylsulfanyl-2-oxo-2H-pyran-3-carbonitrile. It may be employed in the preparation of pyrrolones, which can be used for the synthesis of 1-acyl-3-aryl-3H-pyrrolo[2′,3′:4,5]pyrimido[6,1-b]benzothiazol-6-ium-2-olates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An interaction of 2-thiazoleacetonitriles with N-(2-chloroacetyl) anthranilic acid ester.
Resnyanska EV, et al.
Heterocycles, 63(4), 797-807 (2004)
Carbanion induced synthesis of pyrido [2, 1-b] benzothiazoles through ring transformation reactions.
Ram VJ, et al.
J. Chem. Res. (M), 2001(10), 421-423 (2001)
Synthesis of pyrrolo [2, 3-c] 2, 7-naphthyridine derivatives by cascade heterocyclization reaction of 2-amino-4-cyanomethyl-6-dialkylamino-3, 5-pyridinedicarbonitriles.
Tverdokhlebov AV, et al.
Tetrahedron, 60(27), 5777-5783 (2004)
A simple synthesis of 1-acyl-3-aryl-3H-pyrrolo [2', 3': 4, 5] pyrimido [6, 1-b] benzothiazol-6-ium-2-olates: Betainic derivatives of a novel heterocyclic system.
Resnyanskaya EV, et al.
Synthesis, 18, 2717-2724 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service