Skip to Content
Merck
All Photos(1)

Key Documents

288373

Sigma-Aldrich

2-Amino-6-methylbenzothiazole

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$76.70

$76.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$76.70

About This Item

Empirical Formula (Hill Notation):
C8H8N2S
CAS Number:
Molecular Weight:
164.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

140-142 °C (lit.)

SMILES string

Cc1ccc2nc(N)sc2c1

InChI

1S/C8H8N2S/c1-5-2-3-6-7(4-5)11-8(9)10-6/h2-4H,1H3,(H2,9,10)

InChI key

DZWTXWPRWRLHIL-UHFFFAOYSA-N

General description

The anti-tetanus activity of 2-amino-6-methylbenzothiazole, a muscle relactant, was studied[1].

Application

2-Amino-6-methylbenzothiazole was used in the preparation of 2-[(6-methyl-1,3-benzothiazol-2-yl)amino]-N-[2-(substituted phenyl/furan-2-yl)-4-oxo-1,3-thiazolidin-3-yl]nicotinamides[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Navin B Patel et al.
Scientia pharmaceutica, 78(4), 753-765 (2010-12-24)
The title compounds 6aâj, 2-[(6-methyl-1,3-benzothiazol-2-yl)amino]-N-[2-(substituted phenyl/furan-2-yl)-4-oxo-1,3-thiazolidin-3-yl]nicotinamides, were prepared from 2-chloropyridine-3-carboxylic acid (1) and 2-amino-6-methyl-benzothiazole (2) by known methods. All the compounds have been established by IR, (1)H NMR, (13)C NMR and elemental analyses. The in vitro antimicrobial screening of the
R A WEBSTER
British journal of pharmacology and chemotherapy, 17, 507-518 (1961-12-01)
The anti-tetanus activity of a number of phenothiazine derivatives and other centrally acting muscle relaxants, such as mephenesin, dicyclopropyl ketoxime, 2-amino-6-methylbenzothiazole and meprobamate, has been determined in rabbits with experimental local tetanus. Structure-activity relationships were obtained for the phenothiazine derivatives

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service