Skip to Content
Merck
All Photos(1)

Key Documents

433780

Sigma-Aldrich

1,3-Didecyl-2-methylimidazolium chloride

96%

Synonym(s):

1,3-didecyl-2-methyl-1H-imidazolium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$60.20

$60.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$60.20

About This Item

Empirical Formula (Hill Notation):
C24H47ClN2
CAS Number:
Molecular Weight:
399.10
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$60.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

powder

mp

82 °C

SMILES string

[Cl-].CCCCCCCCCCn1cc[n+](CCCCCCCCCC)c1C

InChI

1S/C24H47N2.ClH/c1-4-6-8-10-12-14-16-18-20-25-22-23-26(24(25)3)21-19-17-15-13-11-9-7-5-2;/h22-23H,4-21H2,1-3H3;1H/q+1;/p-1

InChI key

GJYWPRKIEORZLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,3-Didecyl-2-methylimidazolium chloride can be used:
  • To prepare a new ionic liquid named 1,3-didecyl-2-methylimidazolium dicyanamide, which is applicable in the water/butan-1-ol separation process.[1]
  • To coat a graphene oxide and magnetite based nanocomposite applicable in the separation of hemin from serum samples.[2]
  • As a pore directing agent in the synthesis of mesoporous hectorites for drug delivery applications.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of water/butan-1-ol based on activity coefficients at infinite dilution in 1, 3-didecyl-2-methylimidazolium dicyanamide ionic liquid
Wlazlo M, et al.
The Journal of Chemical Thermodynamics, 116, 316-322 (2018)
Mesoporous synthetic hectorites: A versatile layered host with drug delivery application
Joshi GV, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 142(2-3), 542-548 (2011)
Solid phase extraction of hemin from serum of breast cancer patients using an ionic liquid coated Fe3O4/graphene oxide nanocomposite, and its quantitation by using FAAS.
Farzin L, et al.
Microchimica Acta, 183(9), 2623-2631 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service