Skip to Content
Merck
All Photos(1)

Documents

574074

Sigma-Aldrich

1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

97%

Synonym(s):

1,3-Bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene, 1,3-Bis[2,6-bis(1-methylethyl)phenyl]-1H-imidazolium chloride, 2,5-Bis(2,6-diisopropylphenyl)imidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37ClN2
CAS Number:
Molecular Weight:
425.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

278 °C (dec.) (lit.)

SMILES string

[Cl-].CC(C)c1cccc(C(C)C)c1-n2cc[n+](c2)-c3c(cccc3C(C)C)C(C)C

InChI

1S/C27H37N2.ClH/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;/h9-21H,1-8H3;1H/q+1;/p-1

InChI key

AVJBQMXODCVJCJ-UHFFFAOYSA-M

Application

Used with palladium acetate to generate an N-heterocyclic carbene catalyst for carbonylative cross-coupling of pyridyl halides with aryl boronic acids.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 63, 682-682 (2007)

Articles

We are proud to offer a number of products used in catalytic amidation technology.

Emerging class of privileged ligands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service