Skip to Content
Merck
All Photos(1)

Key Documents

432253

Sigma-Aldrich

Teflic acid

Synonym(s):

Tellurium fluoride hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF5OTe
CAS Number:
Molecular Weight:
239.60
MDL number:
UNSPSC Code:
12352300

vapor pressure

0.24 atm ( 25 °C)

bp

~60 °C (lit.)

mp

~40 °C (lit.)

density

2.5 g/mL at 25 °C (lit.)

SMILES string

O[Te](F)(F)(F)(F)F

Application

Nearly ideal noncoordinating ligand with kinetic and oxidative stability. Precursor to teflate-stabilized rhenium(VII) compounds.

Packaging

Packaged under vacuum in breakseal ampules

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seppelt, K.
Angewandte Chemie (International Edition in English), 21, 877-877 (1982)
William J. Casteel et al.
Inorganic chemistry, 35(25), 7279-7288 (1996-12-04)
The pentafluorooxotellurate compound ReO(2)(OTeF(5))(3) has been synthesized from the reaction of ReO(2)F(3) with B(OTeF(5))(3) and structurally characterized in solution by (19)F and (125)Te NMR spectroscopy and in the solid state by Raman spectroscopy. The NMR and vibrational spectroscopic findings are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service