Standard warranty of one year from date of purchase.
Z148512
Aldrich® corrosive lecture-bottle gas regulator
delivery pressure 1-50 psig, CGA Inlet size 180
Synonym(s):
Corrosive Gas Regulator, Lecture-Bottle Regulator
Select a Size
$3,460.00
Estimated to ship on24 March 2025
Select a Size
About This Item
$3,460.00
Estimated to ship on24 March 2025
Recommended Products
delivery pressure measuring range
1-50 psig
CGA Inlet size
180
General description
- Single-stage
- CGA inlet connection, needle valve outlet with ¼ in. NPTM connection
- Max. inlet pressure: 0-4000 psig
- Temp. range: -40 to 140°F
- 316 Stainless Steel body 1 ½ in. gauge face, PTFE seals and inner friction sleeves
- NPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure; order bonnet-vent tubing connectors separately
- Use with "T"-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipment
- Use for high-purity applications
Lecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Customers Also Viewed
-
What is the warranty on this product?
1 answer-
Helpful?
-