Skip to Content
Merck
All Photos(2)

Key Documents

431257

Sigma-Aldrich

Polyisoprene, cis

average Mw ~38,000 by GPC, made from natural rubber

Synonym(s):

Poly(1-methyl-1-butene-1,4-diyl)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$120.00

$120.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$120.00

About This Item

Linear Formula:
[CH2CH=C(CH3)CH2]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$120.00


Available to ship on22 April 2025Details


Request a Bulk Order

description

unsaturation 98 mole %

form

viscous liquid

mol wt

average Mw ~38,000 by GPC

viscosity

350 poise(37 °C, Brookfield)(lit.)

density

0.92 g/mL at 25 °C

SMILES string

CC(=C)C=C

InChI

1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3

InChI key

RRHGJUQNOFWUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-fugitive, reactive plasticizer, rheology modifier for lubricants, reactive binder.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. I am wondering if the polyisoprene is made from resins of balata or gutta-percha, because it is mentioned that it is made from natural rubber, or is its whole production synthetic?

    1 answer
    1. This product is indicated as being made from natural rubber. Natural rubber is generally sourced from the latex of different trees or plants. Unfortunately, there are no specific details on the type of tree or plant that the natural rubber is obtained from. Please be aware that this product is intended for R&D use, and specific details on manufacturing are not available for this level of product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service