Skip to Content
Merck
All Photos(2)

Key Documents

182168

Sigma-Aldrich

Polyisoprene, trans

pellets, 99+% trans-1,4

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$396.00

$396.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$396.00

About This Item

Linear Formula:
[CH2CH=C(CH3)CH2]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$396.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

form

pellets

transition temp

Tg −68 °C
Tm 60 °C

density

0.904 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C=C

InChI

1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3

InChI key

RRHGJUQNOFWUDK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service