Skip to Content
Merck
All Photos(1)

Key Documents

429503

Sigma-Aldrich

1,1,1,5,5,6,6,6-Octafluoro-2,4-hexanedione

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2F5COCH2COCF3
CAS Number:
Molecular Weight:
258.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
90%

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.3269 (lit.)

bp

85-86 °C (lit.)

density

1.538 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

FC(F)(F)C(=O)CC(=O)C(F)(F)C(F)(F)F

InChI

1S/C6H2F8O2/c7-4(8,6(12,13)14)2(15)1-3(16)5(9,10)11/h1H2

InChI key

MGKBKOFWQWACLM-UHFFFAOYSA-N

Application

1,1,1,5,5,6,6,6-Octafluoro-2,4-hexanedione may be used in the extraction of transition metal ions from water into fluorous solvents.[1] It may be used as a chelating agent for chemical vapor deposition (CVD) of barium fluoride thin films.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal Extraction from Water and Organic Solvents into Fluorous Solvents by Fluorinated β-Diketone and Its Application to the Colorimetric Analysis of Metal Ions.
Nakashima K, et al.
Analytical Sciences, 25(1), 77-77 (2009)
Novel volatile barium β-diketone chelates for chemical vapor deposition of barium fluoride thin films.
Sato H and Sugawara S.
Inorganic Chemistry, 32(10), 1941-1945 (1993)
Carl L. Yaws
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 89-89 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service