Skip to Content
Merck
All Photos(1)

Key Documents

533092

Sigma-Aldrich

4,4-Difluoro-1-phenyl-1,3-butanedione

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$264.00

$264.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$264.00

About This Item

Linear Formula:
C6H5COCH2COCHF2
CAS Number:
Molecular Weight:
198.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$264.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

47-51 °C (lit.)

functional group

fluoro
ketone
phenyl

SMILES string

FC(F)C(=O)CC(=O)c1ccccc1

InChI

1S/C10H8F2O2/c11-10(12)9(14)6-8(13)7-4-2-1-3-5-7/h1-5,10H,6H2

InChI key

JTIPWONXTZMDOK-UHFFFAOYSA-N

General description

4,4-Difluoro-1-phenyl-1,3-butanedione (DFPB) is a fluorinated β-diketone ligand that can be prepared by reacting acetophenone with methyl-difluoroacetate.[1]

Application

4,4-Difluoro-1-phenyl-1,3-butanedione (DPBD) may be used as a ligand to form the following metal complexes: [1]
  • Eu(DPBD)3·H2O
  • Eu(DPBD)3·biq (biq= 2,2-biquinoline)
  • Eu(DPBD)3·phen (phen= 1,10-phenanthroline)
  • Eu(DPBD)3·neo (neo= neocuproine)
  • Eu(DPBD)3·bipy (bipy= 2,2-bipyridyl)
  • Gd(DPBD)3·(H2O)2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, photoluminescent features and intramolecular energy transfer mechanism of europium (III) complexes with fluorinate ?-diketone ligand and auxiliary ligands.
Bala M, et al.
Journal of Fluorine Chemistry, 178, 6-13 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service