Skip to Content
Merck
All Photos(3)

Key Documents

415219

Sigma-Aldrich

Strontium hydroxide octahydrate

95%

Synonym(s):

Strontium dihydroxide octahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$86.80
1 KG
$125.00

$86.80


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$86.80
1 KG
$125.00

About This Item

Linear Formula:
Sr(OH)2 · 8H2O
CAS Number:
Molecular Weight:
265.76
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

$86.80


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

95%

form

crystals and lumps

impurities

SrCO3, trace

density

1.9 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O[Sr]O

InChI

1S/10H2O.Sr/h10*1H2;/q;;;;;;;;;;+2/p-2

InChI key

UJPWWRPNIRRCPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Strontium hydroxideoctahydrate is a white crystalline substance that is commonly used in the production of ceramics, as a raw material for other strontium compounds, and as a chemical reagent. It is also used as a sol-gel precursor to fabricate Sr-based thin films for various catalytic and optoelectronic applications.

Application

Strontium hydroxide octahydrate can be used:
  • As a precursor to synthesize Sr-based nanomaterials by sol-gel hydrothermal method.[1]
  • To fabricate perovskite solar cells with enhanced performance.[2]
  • To synthesize solid base catalysts for solid-liquid interface reactions.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of silver-strontium titanate hybrid nanoparticles by sol-gel-hydrothermal method
Shintaro Ueno, et al.
Nanomaterials, 5, 386-397 (2015)
Synthesis of highly active solid base catalysts of SrO-ZrO2 and SrO-SiO2 by solid?liquid interface reactions of hydrated strontium hydroxide with metal alkoxides
Hiromi Matsuhashi
Catalysis Today, 410, 247-252 (2023)
Graphene/SrTiO3 nanocomposites used as an effective electron-transporting layer for high-performance perovskite solar cells
Chen Wang, et al.
Royal Society of Chemistry Advances, 5, 52041-52047 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service