Skip to Content
Merck
All Photos(2)

Key Documents

415200

Sigma-Aldrich

Strontium peroxide

greener alternative

98%

Synonym(s):

Strontium dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$53.40

$53.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$53.40

About This Item

Linear Formula:
SrO2
CAS Number:
Molecular Weight:
119.62
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$53.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

4.56 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O1O[Sr]1

InChI

1S/O2.Sr/c1-2;/q-2;+2

InChI key

UHCGLDSRFKGERO-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for Molecular Solar Thermal Energy Storage Systems (MOST). Click here for more information.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service