Skip to Content
Merck
All Photos(2)

Key Documents

410284

Sigma-Aldrich

Glycerol propoxylate

average Mn ~266

Synonym(s):

Glycerol propoxylate (1PO/OH)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$208.00

$208.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
1 L
$208.00

About This Item

Linear Formula:
HO(C3H6O)nCH[CH2(OC3H6)nOH]2
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$208.00


Estimated to ship on01 April 2025


Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.3 mmHg ( 20 °C)

form

liquid

mol wt

average Mn ~266

refractive index

n20/D 1.46

solubility

H2O: soluble 1.000 g/L

density

1.09 g/mL at 25 °C

SMILES string

O(C(OCCC(O)CO)OCCC(O)CO)CC(O)CO

InChI

1S/C12H26O9/c13-5-9(16)1-3-19-12(21-8-11(18)7-15)20-4-2-10(17)6-14/h9-18H,1-8H2

InChI key

ZSKSVMDBXFSLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol propoxylate is a triol material with azig-zag structure having a polypropylene glycol unit with three hydroxy end groups. It is a hydrophobic macromonomer widely used in the preparation of crosslinked polymers due to the presence of multiple reaction sites in the branched structure.[1] It can also be used as a plasticizer to impart good elasticity to the materials.[2]

Application

Glycerol propoxylate can be used as a macromonomer to prepare reusablegel oil sorbents for oil spill clean-up.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanical Properties and Thermal Decomposition Behaviors of Hydroxyl-Terminated Polybutadiene/Glycerol Propoxylate Blend and Its Application to Ammonium Nitrate-Based Propellants
Makoto Kohga and Ryoto Togashi
Propellants, Explosives, Pyrotechnics, 46, 1016-1022 (2021)
Oil loving hydrophobic gels made from glycerol propoxylate: Efficient and reusable sorbents for oil spill clean-up
Soner Kizil and Hayal
Journal of Environmental Management, 196, 330-339 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service