Skip to Content
Merck
All Photos(2)

Key Documents

347493

Sigma-Aldrich

(Trimethylsilyl)methacrylate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$260.00
100 G
$779.00

$260.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$260.00
100 G
$779.00

About This Item

Linear Formula:
H2C=C(CH3)CO2Si(CH3)3
CAS Number:
Molecular Weight:
158.27
Beilstein:
1924212
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$260.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

<5 mmHg ( 25 °C)

Quality Level

Assay

98%

form

liquid

contains

500 ppm BHT as inhibitor

refractive index

n20/D 1.415 (lit.)

bp

51-51.5 °C/20 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)O[Si](C)(C)C

InChI

1S/C7H14O2Si/c1-6(2)7(8)9-10(3,4)5/h1H2,2-5H3

InChI key

PGQNYIRJCLTTOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Protected methacrylate monomer which can be deprotected following polymerization.

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Spencer et al.
International journal of pharmaceutics, 588, 119691-119691 (2020-07-30)
Cationic polymers have garnered significant interest for their utility in intracellular drug delivery and gene therapy. However, due to their associated toxicities, novel synthesis approaches must be explored to develop materials that are biocompatible. The novel library of nanoparticles synthesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service