Skip to Content
Merck
All Photos(2)

Key Documents

401714

Sigma-Aldrich

Vinyl propionate

contains <100 ppm monomethyl ether hydroquinone as inhibitor, 98%

Synonym(s):

Vinyl propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$104.00

$104.00


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$104.00

About This Item

Linear Formula:
CH3CH2COOCH=CH2
CAS Number:
Molecular Weight:
100.12
Beilstein:
1741731
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$104.00


Estimated to ship on27 March 2025


Request a Bulk Order

Quality Level

Assay

98%

contains

<100 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.403 (lit.)

bp

94-95 °C (lit.)

mp

−80 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(=O)OC=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h4H,2-3H2,1H3

InChI key

UIWXSTHGICQLQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiao-Feng Li et al.
Journal of agricultural and food chemistry, 67(1), 299-307 (2018-12-19)
Puerarin, an important isoflavonoid from the edible root of Pueraria lobata, shows multiple bioactivities but suffers from low bioavailability. In this study, a new whole-cell catalytic method for acylation modification of puerarin was developed. Among the 12 strains tested, Aspergillus
Jiajia Ding et al.
Carbohydrate polymers, 157, 1785-1793 (2016-12-19)
In this research, cellulose aliphatic esters (CEs) were synthesized efficiently in an N, N-dimethylacetamide/lithium chloride system (DMAc/LiCl) with vinyl esters (VEs, number of carbon atoms ranging from 4 to 12) as acylation reagent and 1,8-diazabicyclo[5,4,0]undec-7-ene (DBU) as catalyst. The structures
Qincao Chen et al.
Food chemistry, 274, 915-924 (2018-10-31)
The formation of and dynamic changes in aroma during white tea processing have not previously been systematically investigated. In this study, advanced comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry was employed to investigate the mechanism of white tea aroma formation. A
Xuan Xin et al.
Microbial cell factories, 17(1), 130-130 (2018-08-24)
Flavonoid glycosides have many beneficial effects on health, but these bioactivities tend to decrease after oral administration owing to their poor lipophilicity. In this study, a facile whole-cell-based method was developed for selective preparation of monoester or diester of troxerutin
Marcela Robles-Machuca et al.
Biotechnology letters, 41(2), 241-252 (2018-12-07)
Ustilago maydis lipase A (UMLA) expressed in Pichia pastoris was compared with Candida antarctica lipase A (CALA) to study its biochemical properties such as thermostability and selectivity. UMLA had similar behavior to its homologue CALA regarding the effect of pH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service