Skip to Content
Merck
All Photos(1)

Key Documents

393770

Sigma-Aldrich

1,3-Benzodithiolylium tetrafluoroborate

98%

Synonym(s):

BDTF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BF4S2
CAS Number:
Molecular Weight:
240.05
Beilstein:
4731453
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%

Assay

98%

form

solid

mp

150 °C (dec.) (lit.)

SMILES string

F[B-](F)(F)F.c1ccc2[s+]csc2c1

InChI

1S/C7H5S2.BF4/c1-2-4-7-6(3-1)8-5-9-7;2-1(3,4)5/h1-5H;/q+1;-1

InChI key

CUSWPJQKCZMDPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Benzodithiolylium tetrafluoroborate (BDTF, 1,3-BDYT) is an electrophilic compound.[1] It is a salt of 1,3-benzodithiolylium ion.[2]
1,3-Benzodithiolylium tetrafluoroborate is a carbenium ionic compound.[3] It has been synthesized by reacting 1,3-benzodithiole with trityl tetrafluoroborate.[2] It is widely used as an α-alkylating agent for the enantioselective α-alkylation of aldehydes.[4],[5] The nature of adsorption of 1,3-benzodithiolylium tetrafluoroborate on various carbon nanotubes (CNTs) has been studied.[1] 1,3-Benzodithiolylium tetrafluoroborate reacts with different nucleophilic reagents at the 2-position to form the respective 1,3-benzodithioles.[2]

Application

1,3-Benzodithiolylium tetrafluoroborate may be used in the synthesis of the following:
  • Dibenzotetrathiafulvalene by reacting with 1,8-diazabicyclo[5.4.0]undec-7-ene.[6]
  • Substituted arylcarbenium ions by reacting with boronic derivatives.[3]
  • α-(1,3-benzodithiol-2-ylidene) ketones and 1,3-benzodithiole by reacting with ketones.[7]
  • 2-styryl-1,3-benzodithioles by reacting with styryl-type cobaloximes.[8]
  • 2-(3-indolyl)-1,3-benzodithiolylium tetrafluoroborate by reacting with indole.[9]
  • 2-(1,3-benzodithiol-2-ylidene)-3,5-cyclohexadien-1-ones and 4-(1,3-benzodithiol-2-ylidene)-2,5-cyclohexadien-1-ones by reacting with 2,4- and 2,6-disubstituted phenols.[10]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Facile and Direct Formylation of Organoboron Aromatic Compounds with Benzodithiolylium Tetrafluoroborate.
Petruzziello D, et al
European Journal of Organic Chemistry, 2013(22), 4909-4917 (2013)
1, 3-Benzodithiolylium salts. Synthesis and reactions with nucleophilic reagents.
Nakayama J, et al
Bulletin of the Chemical Society of Japan, 49(12), 3567-3573 (1976)
The synthesis of highly stable o-and p-quinone methides.
Nakayama J, et al
Chemistry Letters (Jpn), 6(7), 789-792 (1977)
A convenient preparation of-oxo ketene dithioacetals.
Nakayama J, et al
Chemistry Letters (Jpn), 16(5), 939-940 (1987)
Adsorption of 1, 3-benzodithiolylium tetrafluoroborate (1, 3-BDYT) on carbon nanotubes.
Park S, et al
The Journal of the Korean Physical Society, 57(1), 1-4 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service