Skip to Content
Merck
All Photos(2)

Documents

388351

Sigma-Aldrich

Triethyl 2-acetylcitrate

99%

Synonym(s):

Acetyl triethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC(COOCH2CH3)(CH2COOCH2CH3)2
CAS Number:
Molecular Weight:
318.32
Beilstein:
1804947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

viscous liquid

refractive index

n20/D 1.439 (lit.)

bp

228-229 °C/100 mmHg (lit.)

density

1.136 g/mL at 25 °C (lit.)

SMILES string

O=C(OCC)C(CC(OCC)=O)(CC(OCC)=O)OC(C)=O

InChI

1S/C14H22O8/c1-5-19-11(16)8-14(22-10(4)15,13(18)21-7-3)9-12(17)20-6-2/h5-9H2,1-4H3

InChI key

WEAPVABOECTMGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service