Skip to Content
Merck
All Photos(1)

Key Documents

393320

Sigma-Aldrich

2,4,6-Cycloheptatriene-1-carbonitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

51-53 °C/1.5 mmHg (lit.)

density

1.017 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

N#CC1C=CC=CC=C1

InChI

1S/C8H7N/c9-7-8-5-3-1-2-4-6-8/h1-6,8H

InChI key

LADCKIXFXIKHQM-UHFFFAOYSA-N

General description

2,4,6-Cycloheptatriene-1-carbonitrile (7-Cyanocycloheptatriene) is a 7-substituted cycloheptatriene. Its synthesis by reacting tropylium cation with cyanide anion has been described.[1] 2,4,6-cycloheptatriene-1-carbonitrile affords toluene or toluonitrile during flash photolysis studies.[2] Its diamagnetic susceptibility has been determined, which is useful for the derivation of its susceptibility exaltation.[3] It reacts with acetyl acetone in the presence of Mn(OAc)3 to form products derived from the norcaradiene structure.[1]

Application

2,4,6-Cycloheptatriene-1-carbonitrile (7-cyanocycloheptatriene ) may be used in the synthesis of 7-carbomethoxycycloheptatriene.[4] It may be used as model compound for EPR[5] and intramolecular proton transfer studies[6] of cycloheptatriene radicals and anions. It can also be used to generate dihydroazocines and hydrooxocins.[7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diamagnetic susceptibilities and susceptibility exaltations of some 7-substituted cycloheptatrienes.
Childs RF and Pikulik I
Canadian Journal of Chemistry, 55(2), 259-265 (1977)
Magnetic Resonance in Chemistry, 28, 364-364 (1990)
The Flash Photolysis of 1, 3, 5-Cycloheptatriene and Some 7-Substituted Derivatives.
Thrush BA and Zwolenik JJ
Bull. Soc. Chim. Belg., 71(11-12), 642-650 (1962)
The 7-Cycloheptatrienylmethyl/7-Norcaradienylmethyl Cation System.
Pikulik I and Childs RF
Canadian Journal of Chemistry, 53(12), 1818-1828 (1975)
Tetrahedron Letters, 28, 2513-2513 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service