Skip to Content
Merck
All Photos(1)

Key Documents

392499

Sigma-Aldrich

1-Iodo-3,4-dimethylbenzene

99%

Synonym(s):

4-Iodo-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3I
CAS Number:
Molecular Weight:
232.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.603 (lit.)

bp

106-108 °C/13 mmHg (lit.)

density

1.633 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(I)cc1C

InChI

1S/C8H9I/c1-6-3-4-8(9)5-7(6)2/h3-5H,1-2H3

InChI key

CSFRCLYFVINMBZ-UHFFFAOYSA-N

General description

1-Iodo-3,4-dimethylbenzene (4-iodo-o-xylene, 3,4-dimethyliodobenzene) is an aryl halide. Its synthesis, NMR and IR spectra has been reported.[1] Its nitration reaction[2] and copper catalyzed trifluoromethylation reaction has been studied.[3]

Application

1-Iodo-3,4-dimethylbenzene (4-iodo-o-xylene, 3,4-dimethyliodobenzene) may be used in the synthesis of the following:
  • 3-arylacrylamide[4]
  • 3,3-diaryl substituted acrylamide[4]
  • 3-arylpropylamine[4]
  • di-tert-butyl 1-(3,4-dimethylphenyl)hydrazine-1,2-dicarboxylate[5]
  • 2,3,7,8-tetramethyldibenzofuran[6]
  • N-(3-(3,4-Dimethylphenyl)-2-phthalimidopropionyl)-2-methylthioaniline[7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Martyn Inman et al.
The Journal of organic chemistry, 77(3), 1217-1232 (2011-10-08)
In a new variation on the Fischer indole synthesis, readily available haloarenes are converted into a wide range of indoles in just two steps by halogen-magnesium exchange and quenching with di-tert-butyl azodicarboxylate, followed by reaction with aldehydes or ketones under
ipso-Nitration of 4-iodo-o-xylene.
Zweig A, et al.
The Journal of Organic Chemistry, 42(25), 4049-4052 (1977)
Synthesis of 2, 3, 7, 8-tetramethyldibenzofuran.
Mazere IV, et al.
Chemistry of Heterocyclic Compounds, 8(12), 1446-1448 (1972)
Towards a practical and efficient copper-catalyzed trifluoromethylation of aryl halides.
Schareina T, et al.
Topics in Catalysis, 55(7-10), 426-431 (2012)
Dmitry Shabashov et al.
Journal of the American Chemical Society, 132(11), 3965-3972 (2010-02-24)
We have developed a method for auxiliary-directed, palladium-catalyzed beta-arylation and alkylation of sp(3) and sp(2) C-H bonds in carboxylic acid derivatives. The method employs a carboxylic acid 2-methylthioaniline- or 8-aminoquinoline amide substrate, aryl or alkyl iodide coupling partner, palladium acetate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service