Skip to Content
Merck
All Photos(2)

Key Documents

521140

Sigma-Aldrich

3-Ethynyltoluene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$50.80
5 G
$207.00

$50.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$50.80
5 G
$207.00

About This Item

Linear Formula:
Ch3C6H4C≡CH
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$50.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5440 (lit.)

bp

170-175 °C (lit.)

density

0.900 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)C#C

InChI

1S/C9H8/c1-3-9-6-4-5-8(2)7-9/h1,4-7H,2H3

InChI key

RENYIDZOAFFNHC-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erzsébet Mernyák et al.
The Journal of steroid biochemistry and molecular biology, 150, 123-134 (2015-04-08)
Novel 16-triazoles in the 13α-estrone series were synthesized via Cu(I)-catalyzed azide-alkyne cycloaddition of the two diastereomeric (on C-16 and on C-17) 16-azido-13α-estra-1,3,5(10)-trien-17-ol 3-benzyl ethers with substituted phenylacetylenes. The new heterocyclic derivatives were evaluated in vitro by means of MTT assays
Krishna Bahadur Somai Magar et al.
Molecular diversity, 17(4), 679-691 (2013-08-03)
The rhodium(II)-catalyzed reactions of cyclic diazo compounds derived from barbituric acid or thiobarbituric acid with arylacetylenes and styrenes were examined. These reactions provide a rapid synthetic route to the preparation of a variety of novel and diverse furo [2,3-d pyrimidine-2,4-diones
Microwave-assisted palladium-catalyzed highly regio-and stereoselective head to head dimerization of terminal aryl alkynes in water.
Buxaderas E, et al.
Royal Society of Chemistry Advances, 4(87), 46508-46512 (2014)
Yasuhito Suzuki et al.
Forensic science international, 278, 228-239 (2017-08-02)
Various autopsy findings are used for the diagnosis of burning death, including inhalation injury, soot in the respiratory tract, vital reaction, blood carboxyhemoglobin (COHb), and blood cyanide. However, not all findings are necessarily complete, and autopsy diagnosis can be difficult
Hanfeng Jin et al.
Rapid communications in mass spectrometry : RCM, 34(21), e8899-e8899 (2020-07-18)
Aromatic hydrocarbons play an important role in the formation and growth of polycyclic aromatic hydrocarbon (PAH) and soot particles. Measurements of their absolute photoionization cross-sections (PICSs), that benefit the quantitative investigation of mass spectrometry, are still lacking, however. PICSs of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service