Skip to Content
Merck
All Photos(1)

Key Documents

391042

Sigma-Aldrich

(Chloromethyl)triethoxysilane

96%

Synonym(s):

(Chloromethyl)triethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiCH2Cl
CAS Number:
Molecular Weight:
212.75
Beilstein:
1811785
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.41 (lit.)

bp

173-176 °C (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](CCl)(OCC)OCC

InChI

1S/C7H17ClO3Si/c1-4-9-12(7-8,10-5-2)11-6-3/h4-7H2,1-3H3

InChI key

ZDOBWJOCPDIBRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(Chloromethyl)triethoxysilane is an organotrialkoxy silane based reagent with chloromethyl as the organofunctional group. It can be used as a starting material for the synthesis of silane based polymers.[1][2][3][4]

Application

Corresponding Grignard self-condenses to form polyfunctional carbosilanes.[5]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron Letters, 34, 2111-2111 (1993)
Influence of the alkyl chain length of HSO3-R-MCM-41 on the esterification of glycerol with fatty acids
DIAZ I, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 80(1-3), 33-42 (2005)
Effect of spacer groups on the performance of MCM-41-supported platinum cluster-derived hydrogenation catalysts
Maity N, et al.
J. Catal., 242(2), 332-339 (2006)
Substituent effects on the sol- gel chemistry of organotrialkoxysilanes
Loy DA, et al.
Chemistry of Materials, 12(12), 3624-3632 (2000)
Siloxane functionalized cyclopentadienyl-molybdenum complexes: Synthesis, characterization and catalytic application
Zhao J, et al.
Inorgorganica Chimica Acta, 358(14), 4201-4207 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service