Skip to Content
Merck
All Photos(2)

Key Documents

409243

Sigma-Aldrich

Chlorotriethoxysilane

98%

Synonym(s):

Triethoxychlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$246.00
100 G
$777.00

$246.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$246.00
100 G
$777.00

About This Item

Linear Formula:
(C2H5O)3SiCl
CAS Number:
Molecular Weight:
198.72
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$246.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.3900 (lit.)

bp

156-157 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCO[Si](Cl)(OCC)OCC

InChI

1S/C6H15ClO3Si/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3

InChI key

JEZFASCUIZYYEV-UHFFFAOYSA-N

General description

Chlorotriethoxysilaneis a useful research chemical. It is generally used as a silane coupling agent. It is used as a precursor for the precipitation of silicon.[1][2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-purity alkoxychlorosilanes as new precursors for precipitation of silica
Mirskov RG, et al.
Doklady Chemistry, 421(2), 194-196 (2008)
Joseph A Burg et al.
Nature communications, 8(1), 1019-1019 (2017-10-19)
Hyperconnected network architectures can endow nanomaterials with remarkable mechanical properties that are fundamentally controlled by designing connectivity into the intrinsic molecular structure. For hybrid organic-inorganic nanomaterials, here we show that by using 1,3,5 silyl benzene precursors, the connectivity of a
Xin Wang et al.
International journal of biological macromolecules, 169, 239-250 (2020-12-22)
In this study, SBA-15 was modified by halogen & haloalkanes and later used to immobilize lipases. The hydrolysis activity and the glycerolysis performance of the immobilized lipases was carefully studied. Highest activity of the immobilized Candida antarctica lipase B (CALB)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service