Skip to Content
Merck
All Photos(2)

Key Documents

339881

Sigma-Aldrich

Diamminedinitritoplatinum(II) solution

3.4 wt. % in dilute ammonium hydroxide

Synonym(s):

diamminebis(nitrito-κN) platinum

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$164.00
100 G
$1,160.00

$164.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$164.00
100 G
$1,160.00

About This Item

Linear Formula:
Pt(NH3)2(NO2)2
CAS Number:
Molecular Weight:
321.15
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$164.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

core: platinum
reagent type: catalyst

concentration

3.4 wt. % in dilute ammonium hydroxide

density

1.015 g/mL at 25 °C

SMILES string

N[Pt](N)([N+]([O-])=O)[N+]([O-])=O

InChI

1S/2HNO2.2H3N.Pt/c2*2-1-3;;;/h2*(H,2,3);2*1H3;/q;;;;+2/p-2

InChI key

FAOUENTVTAXLPG-UHFFFAOYSA-L

Application

Diamminedinitritoplatinum(II) is an active precursor, extensively used in the preparation of Pt-based catalysts on supports such as carbon black and graphene nanosheets.[1][2][3] Diamminedinitritoplatinum(II) solution can be used as a platinum source in the synthesis of CoXPt1-X alloy nanowires.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and magnetic properties of fcc Co X Pt 1? X nanowires.
Mallet J, et al.
Applied Physics Letters, 84(19), 3900-3902 (2004)
Highly loaded carbon black supported Pt catalysts for fuel cells.
Kaluza L, et al.
Catalysis Today, 256, 375-383 (2015)
EunJoo Yoo et al.
Nano letters, 9(6), 2255-2259 (2009-05-02)
Graphene nanosheet (GNS) gives rise to an extraordinary modification to the properties of Pt cluster electrocatalysts supported on it. The Pt/GNS electrocatalyst revealed an unusually high activity for methanol oxidation reaction compared to Pt/carbon black catalyst. The Pt/GNS electrocatalyst also
Synthesis of sugar alcohols by hydrolytic hydrogenation of cellulose over supported metal catalysts.
Kobayashi H, et al.
Green Chemistry, 13(2), 326-333 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service