377368
Sodium metatungstate
in H2O
Synonym(s):
Sodium polytungstate solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
3Na2WO4·9WO3
CAS Number:
Molecular Weight:
2968.00
MDL number:
UNSPSC Code:
12000000
Recommended Products
composition
W, 55%
concentration
in H2O
refractive index
n20/D 1.5555
density
2.82 g/mL at 25 °C
SMILES string
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service