Skip to Content
Merck
All Photos(1)

Key Documents

375632

Sigma-Aldrich

(1R)-10-Camphorsulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3S
CAS Number:
Molecular Weight:
231.31
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −22°, c = 1 in methanol

mp

133-135 °C (lit.)

functional group

ketone

SMILES string

CC1(C)[C@H]2CC[C@]1(CS(N)(=O)=O)C(=O)C2

InChI

1S/C10H17NO3S/c1-9(2)7-3-4-10(9,8(12)5-7)6-15(11,13)14/h7H,3-6H2,1-2H3,(H2,11,13,14)/t7-,10-/m0/s1

InChI key

SBLUNABTQYDFJM-XVKPBYJWSA-N

Other Notes

Sign of optical rotation varies with solvent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service