Skip to Content
Merck
All Photos(1)

Key Documents

318361

Sigma-Aldrich

(1S)-(−)-Camphorsulfonylimine

99%

Synonym(s):

(1S)-(−)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.01,5]dec-4-ene 3,3-dioxide, (3aS)-(−)-4,5,6,7-Tetrahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole-2,2-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$67.90

$67.90


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$67.90

About This Item

Empirical Formula (Hill Notation):
C10H15NO2S
CAS Number:
Molecular Weight:
213.30
Beilstein:
85296
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$67.90


0 left in stock

Request a Bulk Order

Assay

99%

optical activity

[α]19/D −34°, c = 1 in chloroform

mp

228-230 °C (lit.)

SMILES string

CC1(C)C2CC[C@]13CS(=O)(=O)N=C3C2

InChI

1S/C10H15NO2S/c1-9(2)7-3-4-10(9)6-14(12,13)11-8(10)5-7/h7H,3-6H2,1-2H3/t7?,10-/m1/s1

InChI key

ZAHOEBNYVSWBBW-OMNKOJBGSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service