Skip to Content
Merck
All Photos(4)

Key Documents

372773

Sigma-Aldrich

Poly(ethylene oxide)

average MV 400,000 (nominal), powder, hydroxyl, BHT as inhibitor

Synonym(s):

Polyethylene oxide, PEO

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$83.20
250 G
$303.00
500 G
$526.00

$83.20


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$83.20
250 G
$303.00
500 G
$526.00

About This Item

Linear Formula:
(-CH2CH2O-)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

$83.20


Available to ship on22 April 2025Details


Request a Bulk Order

Product Name

Poly(ethylene oxide), average Mv 400,000 (nominal), powder

form

powder

mol wt

average Mv 400,000 (nominal)

contains

200-500 ppm BHT as inhibitor

viscosity

2,250-4,500 cP, 5 % in H2O(25 °C, Brookfield)(lit.)

transition temp

Tm 65 °C

Ω-end

hydroxyl

α-end

hydroxyl

application(s)

battery manufacturing

SMILES string

[H]OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene oxide) (PEO) is a higher molecular weight (Mv 400,000) thermoplastic water soluble polymer that is formed by the polymerization of ethylene oxide.[1]

Application

PEO can be used in applications such as biomedical, energy and surface functionalization of nanoparticles.[2][3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(Ethylene Oxide) (2012)
Fabrication and properties of composites of poly (ethylene oxide) and functionalized carbon nanotubes
Geng HZ, et al.
Advanced Materials, 14(19), 1387-1390 (2002)
Poly (ethylene oxide)-based electrolytes for lithium-ion batteries
Xue Z, et al.
Journal of Material Chemistry A, 3(38), 19218-19253 (2015)
Polycaprolactone-b-poly (ethylene oxide) copolymer micelles as a delivery vehicle for dihydrotestosterone
Allen C, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 63(3), 275-286 (2000)
Alexander Yang et al.
Biophysical journal, 112(7), 1462-1471 (2017-04-14)
Many species of bacteria can spread over a moist surface via a particular form of collective motion known as "surface swarming". This form of motility is typically studied by inoculating bacteria on a gel formed by 0.4-1.5% agar, which contains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service