Skip to Content
Merck
All Photos(1)

Key Documents

8.18892

Sigma-Aldrich

Polyethylene glycol

35000, (stabilised with 2-tert-butyl-4-methoxyphenol) for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 35000, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 35000, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$92.00
5 KG
$365.00

$92.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$92.00
5 KG
$365.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22

$92.00


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

>320 °C

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

60-65 °C

transition temp

flash point 260 °C

solubility

500 g/L

density

1.2 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 35000 (PEG 35000) is a hydrophilic polyether compound that can be used to synthesize:
  • Hierarchically structured silica monoliths for the fabrication of micro-bioreactors.[1]
  • Micro-nanostructured PEG-DA (PEG-diacrylates) hydrogels for biomedical applications.[2]

It can also be used as:
  • A template to prepare micro/mesoporous structure of silica by sol-gel process.[3]
  • A stabilizing agent in the synthesis of metal nanoparticles.[4]

Analysis Note

Hydroxyl value: 3 - 4
Melting range (lower value): ≥ 62 °C
Melting range (upper value): ≤ 67 °C
Average molecular mass: 28000 - 38000
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the molecular weight?

    1 answer
    1. The average molecular mass of this product is 28000-38000 as mentioned in the "Description Section" of the product detail page. The exact molecular mass will be provided per batch as a result on the Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service