Skip to Content
Merck
All Photos(2)

Key Documents

338184

Sigma-Aldrich

Nickel(II) 2-ethylhexanoate

78% (in 2-Ethylhexanoic acid)

Synonym(s):

2-Ethylhexanoic acid nickel(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CO2]2Ni
Molecular Weight:
345.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

78% (in 2-Ethylhexanoic acid)

form

semisolid
viscous liquid

composition

Ni, 15%

reaction suitability

core: nickel

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)C(=O)O[Ni]OC(=O)C(CC)CCCC

InChI

1S/2C8H16O2.Ni/c2*1-3-5-6-7(4-2)8(9)10;/h2*7H,3-6H2,1-2H3,(H,9,10);/q;;+2/p-2

InChI key

UVPKUTPZWFHAHY-UHFFFAOYSA-L

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.9 °F - closed cup

Flash Point(C)

68.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service