Skip to Content
Merck
All Photos(1)

Key Documents

246050

Sigma-Aldrich

1,5-Cyclooctadiene

purified by redistillation, ≥99%

Synonym(s):

1,5-COD, cis-1,5-Cyclooctadiene, COD

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$102.00
800 ML
$514.00

$102.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$102.00
800 ML
$514.00

About This Item

Empirical Formula (Hill Notation):
C8H12
CAS Number:
Molecular Weight:
108.18
Beilstein:
2036542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$102.00


Available to ship on22 April 2025Details


Request a Bulk Order

vapor pressure

25.8 mmHg ( 37.7 °C)
6.8 mmHg ( 25 °C)

Quality Level

Assay

≥99%

form

liquid

autoignition temp.

431 °F

purified by

redistillation

does not contain

stabilizer

refractive index

n20/D 1.493 (lit.)

bp

149-150 °C (lit.)

mp

−69 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

SMILES string

C1CC=CCCC=C1

InChI

1S/C8H12/c1-2-4-6-8-7-5-3-1/h1-2,7-8H,3-6H2/b2-1-,8-7-

InChI key

VYXHVRARDIDEHS-QGTKBVGQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,5-Cyclooctadiene was used as a catalyst in the synthesis of aminoindene derivatives by the [3+2] annulation of ketimines with internal and terminal alkynes[1].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Midori Nagamoto et al.
Chemical communications (Cambridge, England), 50(47), 6274-6277 (2014-05-07)
[3 + 2] Annulation of ketimines with internal and terminal alkynes proceeded via C-H activation to give aminoindene derivatives in high yields, which is catalyzed by a cationic iridium complex coordinated with 1,5-cyclooctadiene (cod).
Eleonora Cavallari et al.
The journal of physical chemistry. B, 119(31), 10035-10041 (2015-07-15)
Hyperpolarization of (13)C carboxylate signals of metabolically relevant molecules, such as acetate and pyruvate, was recently obtained by means of ParaHydrogen Induced Polarization by Side Arm Hydrogenation (PHIP-SAH). This method relies on functionalization of the carboxylic acid with an unsaturated
Adrian Tlahuext-Aca et al.
Dalton transactions (Cambridge, England : 2003), 43(42), 15997-16005 (2014-09-19)
Ni(0)-catalyzed dehydrogenation of benzylic-type imines was performed to yield asymmetrical tetra-substituted imidazoles and 2-imidazolines. This was achieved with a single operational step while maintaining good selectivity and atom economy. The catalytic system shows low to moderate tolerance for fluoro-, trifluoromethyl-

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service