Skip to Content
Merck
All Photos(2)

Key Documents

330736

Sigma-Aldrich

exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$111.00
100 G
$307.00

$111.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$111.00
100 G
$307.00

About This Item

Empirical Formula (Hill Notation):
C8H6O4
CAS Number:
Molecular Weight:
166.13
Beilstein:
10355
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$111.00


Available to ship on22 April 2025Details


Request a Bulk Order

form

solid

Quality Level

mp

118 °C (dec.) (lit.)

SMILES string

O=C1OC(=O)[C@H]2C3OC(C=C3)[C@@H]12

InChI

1S/C8H6O4/c9-7-5-3-1-2-4(11-3)6(5)8(10)12-7/h1-6H/t3-,4+,5-,6+

InChI key

QQYNRBAAQFZCLF-FBXFSONDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alysia Cox et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 142, 70-82 (2019-06-10)
Nanoparticles may provide a viable way for neuroprotective drugs to cross the blood-brain barrier (BBB), which limits the passage of most drugs from the peripheral circulation to the brain. Heterotelechelic polymer prodrugs comprising a neuroprotective model drug (adenosine) and a
Chao Wu et al.
Advanced materials (Deerfield Beach, Fla.), 32(21), e2000499-e2000499 (2020-04-07)
Flexible dielectrics operable under simultaneous electric and thermal extremes are critical to advanced electronics for ultrahigh densities and/or harsh conditions. However, conventional high-performance polymer dielectrics generally have conjugated aromatic backbones, leading to limited bandgaps and hence high conduction loss and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service