Skip to Content
Merck
All Photos(1)

Key Documents

45347

Sigma-Aldrich

Epoxy embedding medium, hardener MNA

≥95.0%

Synonym(s):

Methyl-5-norbornene-2,3-dicarboxylic anhydride, Epon MNA substitute, MNA, Methylnorbornene-2,3-dicarboxylic acid anhydride, Methyl nadic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$165.00
1 L
$276.00

$165.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$165.00
1 L
$276.00

About This Item

Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
Beilstein:
162395
EC Number:
UNSPSC Code:
41102904
PubChem Substance ID:
NACRES:
MA.02

$165.00


Estimated to ship on12 April 2025


Request a Bulk Order

biological source

synthetic

Quality Level

vapor density

6.1 (20 °C, vs air)

vapor pressure

5 mmHg ( 120 °C)

Assay

≥95.0%

refractive index

n20/D 1.506 (lit.)
n20/D 1.507

density

1.232 g/mL at 25 °C (lit.)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[H][C@]12C3CC(C(C)=C3)[C@@]1([H])C(=O)OC2=O

InChI

1S/C10H10O3/c1-10-6-3-2-5(4-6)7(10)8(11)13-9(10)12/h2-3,5-7H,4H2,1H3/t5-,6+,7+,10+/m0/s1

InChI key

LTVUCOSIZFEASK-MPXCPUAZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Epoxy embedding medium is generally used for biological tissues during study by electron microscopy. Usually, it consists of three components; Epon 812 resin, an anhydride hardener and an accelerator.[1]
additional information see 45359 Epoxy Embedding Medium kit

Application

It was used during transmission electron microscopy done for the analysis of epithelial cells; during a study performed to understand the epithelial cell growth and stratification in minimal culture conditions.[2]

Legal Information

Epon is a trademark of Hexion, Inc.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

275.0 °F

Flash Point(C)

135 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long Xiao et al.
Oxidative medicine and cellular longevity, 2020, 6797154-6797154 (2020-06-23)
A reduction in bone mass around an implant is the main cause of implant loosening, especially in postmenopausal osteoporosis patients. In osteoporosis, excessive oxidative stress, resulting in osteoblast apoptosis, largely contributes to abnormal bone remodeling. Melatonin (MT) synthesized by the
SECTION STAINING FOR ELECTRON MICROSCOPY Incompatibility of Methyl Nadic Anhydride with Permanganates.
Reedy, Michael K.
The Journal of Cell Biology, 26.1, 309-311 (1965)
Federica Riva et al.
Methods in molecular biology (Clifton, N.J.), 585, 25-43 (2009-11-13)
Biological risk management is required in modern tissue engineering. Particular attention should be paid to the culture medium and the scaffold used. In this perspective, it is important to define minimal culture conditions which allow proper growth and differentiation of
Danijela Petrovic et al.
Experimental and molecular pathology, 104(3), 199-211 (2018-05-05)
In our recent studies we have designed fullerenol/doxorubicin nanocomposite (FNP/DOX) as the new drug nanocarrier. This research has demonstrated that this novel nanocomposite has had better implications on the liver tissue in vivo (Wistar rats treated intraperitoneally), than treatment based
Xiao-Wei Zhang et al.
Autophagy, 16(5), 782-796 (2019-07-10)
Impaired macroautophagy/autophagy is involved in the pathogenesis of hepatic fibrosis. However, how aberrant autophagy promotes fibrosis is far from understood. Here, we aimed to define a previously unrevealed pro-fibrotic mechanism for the stress protein TRIB3 (tribbles pseudokinase 3)-mediated autophagy dysfunction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service