320242
Chloroform
ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Synonym(s):
Methylidyne trichloride, Trichloromethane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12190000
Recommended Products
vapor density
4.1 (vs air)
vapor pressure
160 mmHg ( 20 °C)
product line
ReagentPlus®
Assay
≥99.8%
contains
0.5-1.0% ethanol as stabilizer
refractive index
n20/D 1.445 (lit.)
bp
60.5-61.5 °C (lit.)
60.5-61.5 °C
mp
−63 °C (lit.)
density
1.492 g/mL at 25 °C (lit.)
SMILES string
ClC(Cl)Cl
InChI
1S/CHCl3/c2-1(3)4/h1H
InChI key
HEDRZPFGACZZDS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C Nagelreiter et al.
International journal of pharmaceutics, 491(1-2), 162-169 (2015-06-29)
The tape stripping method is a very important tool for dermopharmacokinetic experiments in vitro and the accurate measurement of the removed corneocytes is key for a reliable calculation of a drug's skin penetration behavior. Therefore, various methods to quantify the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service