Skip to Content
Merck
All Photos(3)

Key Documents

308374

Sigma-Aldrich

Lithium thiocyanate hydrate

Synonym(s):

Lithium rhodanide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$194.00
100 G
$557.00

$194.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$194.00
100 G
$557.00

About This Item

Linear Formula:
LiSCN · xH2O
CAS Number:
Molecular Weight:
65.02 (anhydrous basis)
Beilstein:
3623307
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$194.00


Available to ship on15 April 2025Details


Request a Bulk Order

Assay

98.0-102.0% dry basis (silver nitrate titration)

Quality Level

form

solid

SMILES string

[H]O[H].[Li]SC#N

InChI

1S/CHNS.Li.H2O/c2-1-3;;/h3H;;1H2/q;+1;/p-1

InChI key

UNTVNJAFDYEXLD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Gorska et al.
Physical chemistry chemical physics : PCCP, 19(11), 7923-7935 (2017-03-07)
This manuscript reports on the novel insight into the development of high voltage carbon/carbon electrochemical capacitors operating in aqueous solutions of alkali metals and ammonium thiocyanates (KSCN, NaSCN, LiSCN, and NH
Paul D Lane et al.
Physical chemistry chemical physics : PCCP, 22(17), 9438-9447 (2020-04-22)
The liquid-liquid transition in supercooled liquid water, predicted to occur around 220 K, is controversial due to the difficulty of studying it caused by competition from ice crystallization (the so-called "no man's land"). In aqueous solutions, it has been predicted
Qianglu Lin et al.
Journal of the American Chemical Society, 139(19), 6644-6653 (2017-04-22)
The use of semiconductor nanocrystal quantum dots (QDs) in optoelectronic devices typically requires postsynthetic chemical surface treatments to enhance electronic coupling between QDs and allow for efficient charge transport in QD films. Despite their importance in solar cells and infrared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service