Skip to Content
Merck
All Photos(1)

Key Documents

302694

Sigma-Aldrich

3-Acetyl-2,5-dimethylfuran

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$52.70

$52.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$52.70

About This Item

Empirical Formula (Hill Notation):
C8H10O2
CAS Number:
Molecular Weight:
138.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$52.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.485 (lit.)

bp

62 °C/0.25 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1cc(C)oc1C

InChI

1S/C8H10O2/c1-5-4-8(6(2)9)7(3)10-5/h4H,1-3H3

InChI key

KBSVBCHYXYXDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Acetyl-2,5-dimethylfuran is a flavouring agent and its specifications were revised.[1]

Application

3-Acetyl-2,5-dimethylfuran was used in synthesis of:
  • furylfulgide, 2-[1-(2, 5-dimethyl-3-furyl) ethylidene]-3-isopropylidene succinic anhydride[2]
  • chalcones[3]
  • 1-(2′,5′-dimethyl-3′-furyl)-3-(aryl)-2-propen-1-one[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochromism of a furylfulgide, 2-[1-(2, 5-dimethyl-3-furyl) ethylidene]-3-isopropylidene succinic anhydride in solvents and polymer films
Yokoyama Y, et al.
Bulletin of the Chemical Society of Japan, 63(6), 1607-1610 (1990)
J Bend et al.
World Health Organization technical report series, (947)(947), 1-225 (2008-06-17)
This report represents the conclusions of a Joint FAO/WHO Expert Committee convened to evaluate the safety of various food additives, including flavouring agents, with a view to recommending acceptable daily intakes (ADIs) and to preparing specifications for identity and purity.
SYNTHESIS AND STUDY OF ANTI-INFLAMMATORY AND ANTIMICROBIAL ACTIVITY OF SOME NEW 1, 3, 5-TRISUBSTITUTED PYRAZOLINES.
Sridhar S, et al.
International Journal of Chemical Sciences, 8(4), 2697-2707 (2010)
Maura Koehle et al.
ChemSusChem, 10(1), 91-98 (2016-12-13)
A four-step catalytic process was developed to produce p-methylstyrene from methylfuran, a biomass-derived species. First, methylfuran was acylated over zeolite H-Beta with acetic anhydride. Second, the acetyl group was reduced to an ethyl group with hydrogen over copper chromite. Third

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service