Recommended Products
Assay
95%
mp
282 °C (dec.) (lit.)
SMILES string
OC(=O)c1cn[nH]c1
InChI
1S/C4H4N2O2/c7-4(8)3-1-5-6-2-3/h1-2H,(H,5,6)(H,7,8)
InChI key
IMBBXSASDSZJSX-UHFFFAOYSA-N
Application
4-Pyrazolecarboxylic acid was used in preparation of ethyl 4-pyrazolecarboxylate.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of protein chemistry, 15(8), 737-742 (1996-11-01)
Improved and efficient techniques have led to an explosive growth in the application of site-directed mutagenesis to the study of enzymes. However, the limited availability of only those 20 amino acids that are translated by the genetic code has prevented
Journal of vascular surgery, 68(5), 1538-1550 (2017-12-16)
Mural angiogenesis and macrophage accumulation are two pathologic hallmarks of abdominal aortic aneurysm (AAA) disease. The heterodimeric transcription factor hypoxia-inducible factor 1 (HIF-1) is an essential regulator of angiogenesis and macrophage function. In this study, we investigated HIF-1 expression and
Life sciences, 231, 116567-116567 (2019-06-17)
Metabotropic glutamate receptor 5 (mGluR5), a member of group I mGluR, exerts its effect via elevation of intracellular Ca2+ level. We here characterized Ca2+ signals in the tsA201 cells transfected with mGluR5 and investigated the role of passages for mGluR5-induced
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service