Skip to Content
Merck
All Photos(1)

Key Documents

644919

Sigma-Aldrich

3-Methylpyrazole-4-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$161.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$161.00

About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$161.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

225-230 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

Cc1n[nH]cc1C(O)=O

InChI

1S/C5H6N2O2/c1-3-4(5(8)9)2-6-7-3/h2H,1H3,(H,6,7)(H,8,9)

InChI key

HLYYXPDTFLUERX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megumi Iwasaki et al.
Biomedical chromatography : BMC, 30(3), 384-389 (2015-07-16)
Elution profiles of kynurenic acid (KYNA) and 7-chlorokynurenic acid (Cl-KYNA) were examined by high-performance liquid chromatography (HPLC) using a triazole-bonded stationary phase column (Cosmosil® HILIC) under isocratic elution of a mobile phase consisting of CH3 CN-aqueous 10 mm ammonium formate between
Naomi Haruta et al.
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 87(10), 641-648 (2011-12-14)
A determination method for 3-methylpyrazole-5-carboxylic acid (MPC), an inhibitor of D-amino acid oxidase (DAAO), in rat plasma was developed by using high-performance liquid chromatography-mass spectrometry (LC-MS). The structural isomer of MPC, 3-methylpyrazole-4-carboxylic acid, was used as an internal standard, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service