Skip to Content
Merck
All Photos(1)

Key Documents

299987

Sigma-Aldrich

Fluorene-4-carboxylic acid

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O2
CAS Number:
Molecular Weight:
210.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

mp

179-182 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cccc2Cc3ccccc3-c12

InChI

1S/C14H10O2/c15-14(16)12-7-3-5-10-8-9-4-1-2-6-11(9)13(10)12/h1-7H,8H2,(H,15,16)

InChI key

WJNBLUOXTBTGMB-UHFFFAOYSA-N

General description

The structures of two phases of fluorene-4-carboxylic acid from room-temperature growths were studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Blackburn et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 6), 1482-1486 (1996-06-15)
The structures of two phases of fluorene-4-carboxylic acid, C14H10O2, from room-temperature growths have been determined. In the alpha phase, O-H...O hydrogen bonding occurs via cyclic dimers about a center of symmetry; in the beta phase, cyclic dimers with O-H...O hydrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service