Skip to Content
Merck
All Photos(1)

Key Documents

636800

Sigma-Aldrich

7-Benzyloxyindole

96%

Synonym(s):

7-(Phenylmethoxy)indole, NSC 92526

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$103.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$103.00

About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

70-74 °C

functional group

phenyl

SMILES string

C(Oc1cccc2cc[nH]c12)c3ccccc3

InChI

1S/C15H13NO/c1-2-5-12(6-3-1)11-17-14-8-4-7-13-9-10-16-15(13)14/h1-10,16H,11H2

InChI key

DIGZMTAFOACVBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of pyrazolo-quinoline derivatives as adenosine receptor antagonists
Reactant for asymmetric total synthesis of diazonamide A
Reactant for preparation of HCV inhibitor
Reactant for preparation of substituted 3-cyanoindoles and 3-(4-pyridinyl)indoles as inhibitors of inosine monophosphate dehydrogenase, with antiproliferative activity on peripheral blood mononuclear cells (PMBC)
Reactant for preparation of [(indolyl)methyl]hydantoin and -thiohydantoin derivatives as treatment of necroptosis
Reactant for preparation of regioisomeric analogs of ED-110, indolocarbazole poisons of human topoisomerase I

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service