Skip to Content
Merck
All Photos(1)

Documents

298433

Sigma-Aldrich

Dichloroborane methyl sulfide complex

in excess methyl sulfide

Synonym(s):

Boron dichloride methyl sulfide complex, Dichloro(dimethyl sulfide)(hydro)boron, Dichloro(dimethyl sulfide)borane, Dichloroborane dimethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S·BHCl2
CAS Number:
Molecular Weight:
144.86
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: reductant

density

1.255 g/mL at 25 °C (lit.)

SMILES string

C[S+](C)[BH-](Cl)Cl

InChI

1S/C2H7BCl2S/c1-6(2)3(4)5/h3H,1-2H3

InChI key

UMDZKXAEAKAIPA-UHFFFAOYSA-N

Related Categories

Application

Reactant for:
  • Hydroboration reactions
  • Pyrolysis reactions
  • Addition reactions
  • Preparation of unsymmetrical borafluorenes
  • Cyclization reactions
  • Dismutation followed by haloboration of azadiboriridine to give boron-nitrogen clusters
  • Markovnikov hydroboration

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service